2-Chlorophenyl cyclobutyl ketone ≥97%

Supplier: Matrix Scientific

086804-5G 086804-1G 086804-2G
102850-730EA 2172.31 USD
102850-730 102850-728 102850-726
2-Chlorophenyl cyclobutyl ketone ≥97%
2-Chlorophenyl cyclobutyl ketone

Hazard: Irritant

Formula: C₁₁H₁₁ClO
MDL Number: MFCD03841273
CAS Number: 898790-91-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR