4-Bromophenyl cyclobutyl ketone ≥97%

Supplier: Matrix Scientific

Synonyms: (4-Bromophenyl)cyclobutyl-methanone

086799-1G 086799-2G 086799-5G
102850-696EA 672.31 USD
102850-696 102850-698 102850-700
4-Bromophenyl cyclobutyl ketone ≥97%
4-Bromophenyl cyclobutyl ketone

Hazard: Irritant

Formula: C₁₁H₁₁BrO
MW: 239.12 g/mol
MDL Number: MFCD03841260
CAS Number: 898790-60-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR