Cyclobutyl-4-methylphenyl ketone ≥97%
Supplier: Matrix Scientific
086792-2G
086792-5G
086792-1G
102850-658EA
2172.31
USD
102850-658
102850-654
102850-656
Cyclobutyl-4-methylphenyl ketone ≥97%
Cyclobutyl-4-methylphenyl ketone
Hazard: Irritant
Formula:
C₁₂H₁₄O |
MDL Number:
MFCD03841247 CAS Number: 53342-39-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...