4-Bromo-2-fluorophenyl cyclopropyl ketone ≥97%

Supplier: Matrix Scientific

086788-2G 086788-5G 086788-1G
102850-630EA 672.31 USD
102850-630 102850-632 102850-634
4-Bromo-2-fluorophenyl cyclopropyl ketone ≥97%
4-Bromo-2-fluorophenyl cyclopropyl ketone

Hazard: Irritant

Formula: C₁₀H₈BrFO
MDL Number: MFCD03841232
CAS Number: 898790-15-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR