2,2-Dimethyl-2'-fluoropropiophenone ≥97%
Supplier: Matrix Scientific
086767-2G
086767-5G
086767-1G
102850-508EA
3450.77
USD
102850-508
102850-504
102850-506
2,2-Dimethyl-2'-fluoropropiophenone ≥97%
2,2-Dimethyl-2'-fluoropropiophenone
Hazard: Irritant
Formula:
C₁₁H₁₃FO |
MDL Number:
MFCD03841189 CAS Number: 560085-36-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...