3'-Chloro-2,2-dimethyl-4'-fluoropropiophenone ≥97%
Supplier: Matrix Scientific
086766-5G
086766-2G
086766-1G
102850-498EA
672.31
USD
102850-498
102850-500
102850-502
3'-Chloro-2,2-dimethyl-4'-fluoropropiophenone ≥97%
3'-Chloro-2,2-dimethyl-4'-fluoropropiophenone
Hazard: Irritant
Formula:
C₁₁H₁₂ClFO |
MDL Number:
MFCD07699556 CAS Number: 898766-33-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...