4'-Bromo-2,2-dimethyl-3'-fluoropropiophenone ≥97%
Supplier: Matrix Scientific
086765-1G
086765-5G
086765-2G
102850-496EA
2172.31
USD
102850-496
102850-492
102850-494
4'-Bromo-2,2-dimethyl-3'-fluoropropiophenone ≥97%
4'-Bromo-2,2-dimethyl-3'-fluoropropiophenone
Hazard: Irritant
Formula:
C₁₁H₁₂BrFO |
MDL Number:
MFCD07699554 CAS Number: 898766-27-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...