2,2,3',5'-Tetramethylpropiophenone ≥97%

Supplier: Matrix Scientific

086764-1G 086764-2G 086764-5G
102850-486EA 672.31 USD
102850-486 102850-488 102850-490
2,2,3',5'-Tetramethylpropiophenone ≥97%
2,2,3',5'-Tetramethylpropiophenone

Hazard: Irritant

Formula: C₁₃H₁₈O
MDL Number: MFCD03841184
CAS Number: 898766-24-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR