2',2,2,6'-Tetramethylpropiophenone ≥97%
Supplier: Matrix Scientific
086763-2G
086763-5G
086763-1G
102850-482EA
1116.92
USD
102850-482
102850-484
102850-480
2',2,2,6'-Tetramethylpropiophenone ≥97%
2',2,2,6'-Tetramethylpropiophenone
Hazard: Irritant
Formula:
C₁₃H₁₈O |
MDL Number:
MFCD03841182 CAS Number: 25115-80-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...