4'-Chloro-2,2-dimethylpropiophenone ≥97%

Supplier: Matrix Scientific

086762-1G 086762-5G
102850-474EA 1009.23 USD
102850-474 102850-478
4'-Chloro-2,2-dimethylpropiophenone ≥97%
4'-Chloro-2,2-dimethylpropiophenone

Hazard: Irritant

Formula: C₁₁H₁₃ClO
CAS Number: 30314-42-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR