4'-Carboethoxy-2,2-dimethylpropiophenone ≥97%
Supplier: Matrix Scientific
086759-5G
086759-1G
102850-460EA
3450.77
USD
102850-460
102850-456
4'-Carboethoxy-2,2-dimethylpropiophenone ≥97%
4'-Carboethoxy-2,2-dimethylpropiophenone
Hazard: Irritant
Formula:
C₁₄H₁₈O₃ |
MDL Number:
MFCD03841172 CAS Number: 62985-52-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...