3',2,2-Trimethylpropiophenone ≥97%
Supplier: Matrix Scientific
Synonyms:
2,2-Dimethyl-1-m-tolylpropan-1-one
086753-1G
086753-2G
086753-5G
102850-420EA
672.31
USD
102850-420
102850-422
102850-424
3',2,2-Trimethylpropiophenone ≥97%
3',2,2-Trimethylpropiophenone
Hazard: Irritant
Formula:
C₁₂H₁₆O MW: 176.26 g/mol |
MDL Number:
MFCD03841163 CAS Number: 50390-49-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...