2',2,2-Trimethylbutyrophenone ≥97%
Supplier: Matrix Scientific
086749-1G
086749-5G
086749-2G
102850-398EA
1116.92
USD
102850-398
102850-400
102850-396
2',2,2-Trimethylbutyrophenone ≥97%
2',2,2-Trimethylbutyrophenone
Hazard: Irritant
Formula:
C₁₃H₁₈O |
MDL Number:
MFCD03841116 CAS Number: 66390-57-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...