4'-Bromo-3,3-dimethyl-3'-fluorobutyrophenone ≥97%
Supplier: Matrix Scientific
086746-5G
086746-2G
086746-1G
102850-378EA
672.31
USD
102850-378
102850-382
102850-380
4'-Bromo-3,3-dimethyl-3'-fluorobutyrophenone ≥97%
4'-Bromo-3,3-dimethyl-3'-fluorobutyrophenone
Hazard: Irritant
Formula:
C₁₂H₁₄BrFO |
MDL Number:
MFCD07699546 CAS Number: 898764-62-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...