2',3,3-Trimethylbutyrophenone ≥97%
Supplier: Matrix Scientific
086738-2G
086738-1G
086738-5G
102850-332EA
1116.92
USD
102850-332
102850-334
102850-330
2',3,3-Trimethylbutyrophenone ≥97%
2',3,3-Trimethylbutyrophenone
Hazard: Irritant
Formula:
C₁₃H₁₈O |
MDL Number:
MFCD03841074 CAS Number: 898785-47-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...