3,3-Dimethylbutyrophenone ≥97%
Supplier: Matrix Scientific
086737-5G
086737-1G
086737-2G
102850-328EA
3450.77
USD
102850-328
102850-324
102850-326
3,3-Dimethylbutyrophenone ≥97%
3,3-Dimethylbutyrophenone
Hazard: Irritant
Formula:
C₁₂H₁₆O |
MDL Number:
MFCD03841073 CAS Number: 31366-07-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...