2-(3-Methoxyphenyl)acetophenone ≥97%

Supplier: Matrix Scientific

086710-5G 086710-1G 086710-2G
102850-166EA 904.62 USD
102850-166 102850-162 102850-164
2-(3-Methoxyphenyl)acetophenone ≥97%
2-(3-Methoxyphenyl)acetophenone

Hazard: Irritant

Formula: C₁₅H₁₄O₂
MDL Number: MFCD00446972
CAS Number: 29955-26-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR