2-(3-Methylphenyl)acetophenone ≥97%
Supplier: Matrix Scientific
086709-1G
086709-5G
086709-2G
102850-156EA
347.69
USD
102850-156
102850-158
102850-160
2-(3-Methylphenyl)acetophenone ≥97%
2-(3-Methylphenyl)acetophenone
Hazard: Irritant
Formula:
C₁₅H₁₄O |
MDL Number:
MFCD02260689 CAS Number: 34403-03-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...