2-Phenyl-3'-(trifluormethyl)acetophenone ≥97%

Supplier: Matrix Scientific

086698-5G 086698-1G 086698-2G
102850-092EA 886.15 USD
102850-092 102850-094 102850-090
2-Phenyl-3'-(trifluormethyl)acetophenone ≥97%
2-Phenyl-3'-(trifluormethyl)acetophenone

Hazard: Irritant

Formula: C₁₅H₁₁F₃O
MDL Number: MFCD02260677
CAS Number: 1533-04-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR