Benzyl-4-methoxyphenyl ketone ≥97%

Supplier: Matrix Scientific

Synonyms: 4'-Methoxy-2-phenylacetophenone1-(4-Methoxyphenyl)-2-phenylethanone

086695-2G 086695-5G 086695-1G
102850-074EA 560 USD
102850-074 102850-076 102850-072
Benzyl-4-methoxyphenyl ketone ≥97%
Benzyl 4-methoxyphenyl ketone

Hazard: Irritant

Formula: C₁₅H₁₄O₂
MW: 226.2700 g/mol
MDL Number: MFCD00017177
CAS Number: 1023-17-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR