Benzyl-3-methoxyphenyl ketone ≥97%
Supplier: Matrix Scientific
086694-2G
086694-1G
086694-5G
102850-068EA
560
USD
102850-068
102850-066
102850-070
Benzyl-3-methoxyphenyl ketone ≥97%
Benzyl 3-methoxyphenyl ketone
Hazard: Irritant
Formula:
C₁₅H₁₄O₂ |
MDL Number:
MFCD02260672 CAS Number: 62381-24-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...