2'-Fluoro-2-phenylacetophenone ≥97%
Supplier: Matrix Scientific
086683-5G
086683-1G
086683-2G
102850-002EA
886.15
USD
102850-002
102850-004
102850-000
2'-Fluoro-2-phenylacetophenone ≥97%
2'-Fluoro-2-phenylacetophenone
Hazard: Irritant
Formula:
C₁₄H₁₁FO |
MDL Number:
MFCD02260662 CAS Number: 77294-82-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...