4-Chloro-1-(2-fluorophenyl)-1-oxobutane ≥97%
Supplier: Matrix Scientific
086678-1G
086678-5G
086678-2G
102849-970EA
964.62
USD
102849-970
102849-974
102849-972
4-Chloro-1-(2-fluorophenyl)-1-oxobutane ≥97%
4-Chloro-1-(2-fluorophenyl)-1-oxobutane
Hazard: Irritant
Formula:
C₁₀H₁₀ClFO |
MDL Number:
MFCD02260553 CAS Number: 2823-19-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...