6-Oxo-6-phenylhexanenitrile ≥97%

Supplier: Matrix Scientific

086668-5G 086668-2G 086668-1G
102849-914EA 904.62 USD
102849-914 102849-912 102849-910
6-Oxo-6-phenylhexanenitrile ≥97%
6-Oxo-6-phenylhexanenitrile

Hazard: Irritant

Formula: C₁₂H₁₃NO
MDL Number: MFCD02260532
CAS Number: 28353-03-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR