5-Oxo-5-phenylvaleronitrile ≥97%
Supplier: Matrix Scientific
086667-5G
086667-1G
086667-2G
102849-908EA
1436.92
USD
102849-908
102849-906
102849-904
5-Oxo-5-phenylvaleronitrile ≥97%
5-Oxo-5-phenylvaleronitrile
Hazard: Irritant
Formula:
C₁₁H₁₁NO |
MDL Number:
MFCD00463744 CAS Number: 10413-00-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...