5-(4-Acetoxybenzoyl)-2-chloropyridine ≥90%

Supplier: Matrix Scientific

086638-1G 086638-2G 086638-5G
102849-746EA 2172.31 USD
102849-746 102849-744 102849-742
5-(4-Acetoxybenzoyl)-2-chloropyridine ≥90%
5-(4-Acetoxybenzoyl)-2-chloropyridine

Hazard: Irritant

Formula: C₁₄H₁₀ClNO₃
MDL Number: MFCD07699489
CAS Number: 898786-50-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR