2-Chloro-4-propionylpyridine ≥97%
Supplier: Matrix Scientific
086633-5G
086633-2G
086633-1G
102849-716EA
3350.77
USD
102849-716
102849-714
102849-712
2-Chloro-4-propionylpyridine ≥97%
2-Chloro-4-propionylpyridine
Hazard: Irritant
Formula:
C₈H₈ClNO Melting Pt: 32…34 °C |
MDL Number:
MFCD07699407 CAS Number: 87121-53-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...