2-(3-Methylbenzoyl)oxazole ≥97%
Supplier: Matrix Scientific
086620-5G
086620-2G
086620-1G
102849-638EA
2172.31
USD
102849-638
102849-636
102849-634
2-(3-Methylbenzoyl)oxazole ≥97%
2-(3-Methylbenzoyl)oxazole
Hazard: Irritant
Formula:
C₁₁H₉NO₂ |
MDL Number:
MFCD07699340 CAS Number: 898759-56-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...