2-(4-Iodobenzoyl)pyridine ≥97%

Supplier: Matrix Scientific

086608-2G 086608-1G 086608-5G
102849-562EA 795.38 USD
102849-562 102849-566 102849-564
2-(4-Iodobenzoyl)pyridine ≥97%
2-(4-Iodobenzoyl)pyridine

Hazard: Irritant

Formula: C₁₂H₈INO
MDL Number: MFCD07699241
CAS Number: 898779-82-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR