2-(4-Chlorobenzoyl)pyridine ≥97%
Supplier: Matrix Scientific
Synonyms:
(4-Chlorophenyl)(pyridin-2-yl)methanone
086604-5G
102849-542EA
207.69
USD
102849-542
2-(4-Chlorobenzoyl)pyridine ≥97%
2-(4-Chlorobenzoyl)pyridine
Hazard: Irritant
Formula:
C₁₂H₈ClNO MW: 217.6500 g/mol |
MDL Number:
MFCD02930888 CAS Number: 6318-51-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...