2-(2-Methoxybenzoyl)pyridine ≥97%
Supplier: Matrix Scientific
086596-2G
086596-5G
086596-1G
102849-494EA
2172.31
USD
102849-494
102849-492
102849-490
2-(2-Methoxybenzoyl)pyridine ≥97%
2-(2-Methoxybenzoyl)pyridine
Hazard: Irritant
Formula:
C₁₃H₁₁NO₂ |
MDL Number:
MFCD07699223 CAS Number: 22945-63-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...