2,2-Dimethylpropyl-2-pyridyl ketone ≥97%

Supplier: Matrix Scientific

086595-1G 086595-2G 086595-5G
102849-486EA 1749.23 USD
102849-486 102849-484 102849-488
2,2-Dimethylpropyl-2-pyridyl ketone ≥97%
2,2-Dimethylpropyl-2-pyridyl ketone

Hazard: Irritant

Formula: C₁₁H₁₅NO
MDL Number: MFCD07699214
CAS Number: 138835-86-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR