Isopropyl-2-pyridyl ketone ≥97%
Supplier: Matrix Scientific
086593-1G
086593-5G
086593-2G
102849-474EA
1136.92
USD
102849-474
102849-472
102849-476
Isopropyl-2-pyridyl ketone ≥97%
Isopropyl-2-pyridyl ketone
Hazard: Irritant
Formula:
C₉H₁₁NO MW: 149.19 g/mol |
MDL Number:
MFCD00778577 CAS Number: 59576-30-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...