Ethyl-4-oxo-4-(3-thienyl)butyrate ≥97%
Supplier: Matrix Scientific
086586-5G
086586-2G
086586-1G
102849-430EA
795.38
USD
102849-430
102849-434
102849-432
Ethyl-4-oxo-4-(3-thienyl)butyrate ≥97%
Ethyl-4-oxo-4-(3-thienyl)butyrate
Hazard: Irritant
Formula:
C₁₀H₁₂O₃S |
MDL Number:
MFCD07699058 CAS Number: 473693-79-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...