2-Chloro-5-[(3-oxocyclohexyl)methyl]pyridine ≥97%
Supplier: Matrix Scientific
086578-2G
086578-5G
086578-1G
102849-386EA
2172.31
USD
102849-386
102849-384
102849-382
2-Chloro-5-[(3-oxocyclohexyl)methyl]pyridine ≥97%
2-Chloro-5-[(3-oxocyclohexyl)methyl]pyridine
Hazard: Irritant
Formula:
C₁₂H₁₄ClNO |
MDL Number:
MFCD03841031 CAS Number: 909421-71-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...