4-(2-Thenoyl)pyridine ≥97%

Supplier: Matrix Scientific

086574-2G 086574-1G 086574-5G
102849-362EA 2172.31 USD
102849-362 102849-360 102849-358
4-(2-Thenoyl)pyridine ≥97%
4-(2-Thenoyl)pyridine

Hazard: Irritant

Formula: C₁₀H₇NOS
MDL Number: MFCD02260531
CAS Number: 21314-80-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR