Ethyl-4-(6-chloropyridin-3-yl)-4-oxobutyrate ≥97%
Supplier: Matrix Scientific
086573-2G
086573-1G
086573-5G
102849-354EA
1136.92
USD
102849-354
102849-352
102849-356
Ethyl-4-(6-chloropyridin-3-yl)-4-oxobutyrate ≥97%
Ethyl-4-(6-chloropyridin-3-yl)-4-oxobutyrate
Hazard: Irritant
Formula:
C₁₁H₁₂ClNO₃ |
MDL Number:
MFCD02260485 CAS Number: 890100-63-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...