2-Chloro-5-(3-ethoxycarbonylbenzoyl)pyridine ≥97%

Supplier: Matrix Scientific

086570-1G 086570-5G 086570-2G
102849-338EA 2172.31 USD
102849-338 102849-336 102849-334
2-Chloro-5-(3-ethoxycarbonylbenzoyl)pyridine ≥97%
2-Chloro-5-(3-ethoxycarbonylbenzoyl)pyridine

Hazard: Irritant

Formula: C₁₅H₁₂ClNO₃
MDL Number: MFCD02260458
CAS Number: 890100-48-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR