4-Bromo-2'-methylbenzophenone ≥97%
Supplier: Matrix Scientific
086564-1G
086564-5G
086564-2G
102849-298EA
376.92
USD
102849-298
102849-302
102849-300
4-Bromo-2'-methylbenzophenone ≥97%
4-Bromo-2'-methylbenzophenone
Hazard: Irritant
Formula:
C₁₄H₁₁BrO Melting Pt: 40…41 °C |
MDL Number:
MFCD09801661 CAS Number: 27428-59-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...