4-Bromo-3'-nitrobenzophenone ≥97%
Supplier: Matrix Scientific
086540-5G
086540-2G
086540-1G
102849-156EA
1381.54
USD
102849-156
102849-154
102849-158
4-Bromo-3'-nitrobenzophenone ≥97%
4-Bromo-3'-nitrobenzophenone
Hazard: Irritant
Formula:
C₁₃H₈BrNO₃ Melting Pt: 105…107 °C |
MDL Number:
MFCD00617167 CAS Number: 62100-13-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...