2-Chloro-4'-nitrobenzophenone ≥97%

Supplier: Matrix Scientific

086538-1G 086538-2G 086538-5G
102849-144EA 847.69 USD
102849-144 102849-142 102849-146
2-Chloro-4'-nitrobenzophenone ≥97%
2-Chloro-4'-nitrobenzophenone

Hazard: Irritant

Formula: C₁₃H₈ClNO₃
MDL Number: MFCD02260377
CAS Number: 77778-73-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR