4'-Bromo-4-iodobenzophenone ≥97%
Supplier: Matrix Scientific
086533-2G
086533-5G
086533-1G
102849-112EA
387.69
USD
102849-112
102849-116
102849-114
4'-Bromo-4-iodobenzophenone ≥97%
4'-Bromo-4-iodobenzophenone
Hazard: Irritant
Formula:
C₁₃H₈BrIO |
MDL Number:
MFCD00510441 CAS Number: 609820-28-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...