3-Fluoro-4'-methoxybenzophenone ≥97%
Supplier: Matrix Scientific
086529-2G
086529-1G
086529-5G
102849-088EA
490.77
USD
102849-088
102849-092
102849-090
3-Fluoro-4'-methoxybenzophenone ≥97%
3-Fluoro-4'-methoxybenzophenone
Hazard: Irritant
Formula:
C₁₄H₁₁FO₂ |
MDL Number:
MFCD01311581 CAS Number: 96719-99-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...