4-Bromo-4'-methoxybenzophenone ≥97%
Supplier: Matrix Scientific
Synonyms:
(4-Bromophenyl)(4-methoxyphenyl)methanone
086528-1G
086528-2G
086528-5G
102849-086EA
684.62
USD
102849-086
102849-084
102849-082
4-Bromo-4'-methoxybenzophenone ≥97%
4-Bromo-4'-methoxybenzophenone
Hazard: Irritant
Formula:
C₁₄H₁₁BrO₂ MW: 291.14 g/mol Melting Pt: 156…159 °C |
MDL Number:
MFCD00228667 CAS Number: 54118-75-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...