4-Carboethoxy-4'-methoxybenzophenone ≥97%
Supplier: Matrix Scientific
086526-5G
086526-1G
086526-2G
102849-074EA
1430.77
USD
102849-074
102849-072
102849-070
4-Carboethoxy-4'-methoxybenzophenone ≥97%
4-Carboethoxy-4'-methoxybenzophenone
Hazard: Irritant
Formula:
C₁₇H₁₆O₄ Melting Pt: 82…84 °C |
MDL Number:
MFCD01311578 CAS Number: 67205-87-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...