3,3'-Dimethoxybenzophenone ≥97%
Supplier: Matrix Scientific
086520-2G
086520-1G
086520-5G
102849-034EA
489.23
USD
102849-034
102849-038
102849-036
3,3'-Dimethoxybenzophenone ≥97%
3,3'-Dimethoxybenzophenone
Hazard: Irritant
Formula:
C₁₅H₁₄O₃ |
CAS Number:
39193-85-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...