2-Chloro-2'-methoxybenzophenone ≥97%
Supplier: Matrix Scientific
086518-1G
086518-5G
086518-2G
102849-024EA
847.69
USD
102849-024
102849-022
102849-026
2-Chloro-2'-methoxybenzophenone ≥97%
2-Chloro-2'-methoxybenzophenone
Hazard: Irritant
Formula:
C₁₄H₁₁ClO₂ Melting Pt: 58…60 °C |
MDL Number:
MFCD07698786 CAS Number: 890098-04-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...