2-Bromo-4'-fluorobenzophenone ≥97%
Supplier: Matrix Scientific
Synonyms:
(2-Bromophenyl)(4-fluorophenyl)methanone
086512-5G
086512-2G
086512-1G
102848-986EA
490.77
USD
102848-986
102848-988
102848-990
2-Bromo-4'-fluorobenzophenone ≥97%
2-Bromo-4'-fluorobenzophenone
Hazard: Irritant
Formula:
C₁₃H₈BrFO MW: 279.12 g/mol |
MDL Number:
MFCD00672033 CAS Number: 573-46-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...