2-Bromo-2'-carboethoxybenzophenone ≥97%
Supplier: Matrix Scientific
086508-2G
086508-1G
086508-5G
102848-962EA
490.77
USD
102848-962
102848-964
102848-966
2-Bromo-2'-carboethoxybenzophenone ≥97%
2-Bromo-2'-carboethoxybenzophenone
Hazard: Irritant
Formula:
C₁₆H₁₃BrO₃ |
MDL Number:
MFCD00672026 CAS Number: 746651-81-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...