2-Bromo-4'-cyanobenzophenone ≥97%

Supplier: Matrix Scientific

086504-1G 086504-2G 086504-5G
102848-940EA 847.69 USD
102848-940 102848-942 102848-938
2-Bromo-4'-cyanobenzophenone ≥97%
2-Bromo-4'-cyanobenzophenone

Hazard: Irritant

Formula: C₁₄H₈BrNO
MDL Number: MFCD00672019
CAS Number: 746651-77-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR